3-{[(4-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049899-500MG 049899-2.5G
101889-496EA 364.62 USD
101889-496 101889-498
3-{[(4-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560301
CAS Number: 1219971-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR