4-{2-[(4-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049896-500MG 049896-2.5G
101889-484EA 364.62 USD
101889-484 101889-486
4-{2-[(4-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(4-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MW: 269.82 g/mol
MDL Number: MFCD13560298
CAS Number: 1220018-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR