3-{[(4-Isopropylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049895-500MG 049895-2.5G
101889-480EA 166.15 USD
101889-480 101889-482
3-{[(4-Isopropylbenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Isopropylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560297
CAS Number: 1219949-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR