4-Isopropylbenzyl-2-(2-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

049894-500MG 049894-2.5G
101889-476EA 364.62 USD
101889-476 101889-478
4-Isopropylbenzyl-2-(2-piperidinyl)ethyl ether hydrochloride
4-Isopropylbenzyl-2-(2-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MW: 297.87 g/mol
MDL Number: MFCD13560296
CAS Number: 1220031-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR