4-Isopropylbenzyl-2-(4-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

049892-500MG 049892-2.5G
101889-468EA 364.62 USD
101889-468 101889-470
4-Isopropylbenzyl-2-(4-piperidinyl)ethyl ether hydrochloride
4-Isopropylbenzyl-2-(4-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MW: 297.87 g/mol
MDL Number: MFCD13560294
CAS Number: 1219971-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR