4-(tert-Butyl)benzyl-3-pyrrolidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
049891-500MG
049891-2.5G
101889-464EA
166.15
USD
101889-464
101889-466
4-(tert-Butyl)benzyl-3-pyrrolidinylmethyl ether hydrochloride
4-(tert-Butyl)benzyl-3-pyrrolidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560293 CAS Number: 1219980-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...