2-(2-{[4-(tert-Butyl)benzyl]oxy}ethyl)piperidine hydrochloride

Supplier: Matrix Scientific

049890-500MG 049890-2.5G
101889-460EA 364.62 USD
101889-460 101889-462
2-(2-{[4-(tert-Butyl)benzyl]oxy}ethyl)piperidine hydrochloride
2-(2-{[4-(tert-Butyl)benzyl]oxy}ethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₃₀ClNO
MW: 311.9 g/mol
MDL Number: MFCD13560292
CAS Number: 1219980-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR