3-{[(4-Chlorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049883-500MG 049883-2.5G
101889-432EA 364.62 USD
101889-432 101889-434
3-{[(4-Chlorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Chlorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560285
CAS Number: 1219949-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR