4-{2-[(4-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049880-500MG 049880-2.5G
101889-420EA 364.62 USD
101889-420 101889-422
4-{2-[(4-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(4-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560282
CAS Number: 1219968-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR