3-{[(4-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049879-500MG 049879-2.5G
101889-416EA 364.62 USD
101889-416 101889-418
3-{[(4-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MDL Number: MFCD13560281
CAS Number: 1219980-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR