2-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049878-500MG 049878-2.5G
101889-412EA 364.62 USD
101889-412 101889-414
2-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClFNO
MDL Number: MFCD13560280
CAS Number: 1219968-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR