4-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049876-500MG 049876-2.5G
101889-404EA 364.62 USD
101889-404 101889-406
4-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(4-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClFNO
MDL Number: MFCD13560278
CAS Number: 1219967-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR