3-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049873-500MG 049873-2.5G
101889-392EA 364.62 USD
101889-392 101889-394
3-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560275
CAS Number: 1219967-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR