4-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049872-500MG 049872-2.5G
101889-388EA 364.62 USD
101889-388 101889-390
4-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MW: 269.82 g/mol
MDL Number: MFCD13560274
CAS Number: 1220031-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR