3-{[(3-Chlorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049871-500MG 049871-2.5G
101889-384EA 364.62 USD
101889-384 101889-386
3-{[(3-Chlorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560273
CAS Number: 1219980-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR