3-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049869-500MG 049869-2.5G
101889-376EA 364.62 USD
101889-376 101889-378
3-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560271
CAS Number: 1219980-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR