4-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049868-500MG
049868-2.5G
101889-372EA
364.62
USD
101889-372
101889-374
4-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO |
MDL Number:
MFCD13560270 CAS Number: 1220031-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...