3-{[(3-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049867-500MG 049867-2.5G
101889-368EA 364.62 USD
101889-368 101889-370
3-{[(3-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(3-Fluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MDL Number: MFCD13560269
CAS Number: 1219967-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR