3-{2-[(3-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049865-500MG 049865-2.5G
101889-360EA 364.62 USD
101889-360 101889-362
3-{2-[(3-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClFNO
MDL Number: MFCD13560267
CAS Number: 1220018-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR