3-{[(2-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049863-500MG 049863-2.5G
101889-352EA 364.62 USD
101889-352 101889-354
3-{[(2-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(2-Methylbenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560265
CAS Number: 1219949-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR