2-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049862-500MG 049862-2.5G
101889-348EA 364.62 USD
101889-348 101889-350
2-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560264
CAS Number: 1219972-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR