3-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049861-500MG 049861-2.5G
101889-344EA 364.62 USD
101889-344 101889-346
3-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560263
CAS Number: 1220028-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR