4-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049860-500MG 049860-2.5G
101889-340EA 166.15 USD
101889-340 101889-342
4-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(2-Methylbenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560262
CAS Number: 1220017-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR