3-{[(2-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049859-500MG 049859-2.5G
101889-336EA 364.62 USD
101889-336 101889-338
3-{[(2-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(2-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560261
CAS Number: 1220036-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR