2-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049858-500MG 049858-2.5G
101889-332EA 364.62 USD
101889-332 101889-334
2-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560260
CAS Number: 1220018-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR