4-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049856-500MG 049856-2.5G
101889-324EA 364.62 USD
101889-324 101889-326
4-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(2-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560258
CAS Number: 1219949-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR