2-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049854-500MG 049854-2.5G
101889-316EA 364.62 USD
101889-316 101889-318
2-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀Cl₃NO
MDL Number: MFCD13560256
CAS Number: 1219971-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR