4-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049852-500MG 049852-2.5G
101889-308EA 364.62 USD
101889-308 101889-310
4-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(2,4-Dichlorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀Cl₃NO
MDL Number: MFCD13560254
CAS Number: 1220017-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR