2-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049850-500MG 049850-2.5G
101889-300EA 364.62 USD
101889-300 101889-302
2-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560252
CAS Number: 1219980-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR