3-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049849-500MG
049849-2.5G
101889-296EA
364.62
USD
101889-296
101889-298
3-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(2-Chlorobenzyl)oxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO |
MDL Number:
MFCD13560251 CAS Number: 1220018-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...