2-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049846-500MG 049846-2.5G
101889-284EA 364.62 USD
101889-284 101889-286
2-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClFNO
MDL Number: MFCD13560248
CAS Number: 1219967-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR