4-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049844-500MG 049844-2.5G
101889-276EA 364.62 USD
101889-276 101889-278
4-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(2-Fluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClFNO
MDL Number: MFCD13560246
CAS Number: 1219980-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR