3-[(Pentyloxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049843-500MG 049843-2.5G
101889-272EA 364.62 USD
101889-272 101889-274
3-[(Pentyloxy)methyl]pyrrolidine hydrochloride
3-[(Pentyloxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO
MDL Number: MFCD13560245
CAS Number: 1220017-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR