4-[2-(Pentyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049840-500MG 049840-2.5G
101889-260EA 364.62 USD
101889-260 101889-262
4-[2-(Pentyloxy)ethyl]piperidine hydrochloride
4-[2-(Pentyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO
MDL Number: MFCD13560242
CAS Number: 1220030-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR