4-[2-(4-Pentenyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049832-500MG 049832-2.5G
101889-228EA 364.62 USD
101889-228 101889-230
4-[2-(4-Pentenyloxy)ethyl]piperidine hydrochloride
4-[2-(4-Pentenyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO
MW: 233.78 g/mol
MDL Number: MFCD13560234
CAS Number: 1220028-68-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR