3-[(Isopentyloxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049831-500MG 049831-2.5G
101889-224EA 364.62 USD
101889-224 101889-226
3-[(Isopentyloxy)methyl]pyrrolidine hydrochloride
3-[(Isopentyloxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO
MDL Number: MFCD13560233
CAS Number: 1220030-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR