2-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049830-500MG 049830-2.5G
101889-220EA 364.62 USD
101889-220 101889-222
2-[2-(Isopentyloxy)ethyl]piperidine hydrochloride
2-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO
MDL Number: MFCD13560232
CAS Number: 1219980-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR