3-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049829-500MG 049829-2.5G
101889-216EA 364.62 USD
101889-216 101889-218
3-[2-(Isopentyloxy)ethyl]piperidine hydrochloride
3-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO
MDL Number: MFCD13560231
CAS Number: 1220030-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR