4-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049828-500MG 049828-2.5G
101889-212EA 364.62 USD
101889-212 101889-214
4-[2-(Isopentyloxy)ethyl]piperidine hydrochloride
4-[2-(Isopentyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO
MDL Number: MFCD13560230
CAS Number: 1220036-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR