3-[2-(Allyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049825-500MG 049825-2.5G
101889-200EA 364.62 USD
101889-200 101889-202
3-[2-(Allyloxy)ethyl]piperidine hydrochloride
3-[2-(Allyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD13560227
CAS Number: 1220036-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR