3-[(2-Propynyloxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
Synonyms:
3-((Prop-2-yn-1-yloxy)methyl)pyrrolidine hydrochloride
049823-500MG
049823-2.5G
101889-192EA
384.62
USD
101889-192
101889-194
3-[(2-Propynyloxy)methyl]pyrrolidine hydrochloride
3-[(2-Propynyloxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₄ClNO MW: 175.66 g/mol |
MDL Number:
MFCD13560225 CAS Number: 1220036-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...