2-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049822-500MG 049822-2.5G
101889-188EA 364.62 USD
101889-188 101889-190
2-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride
2-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO
MDL Number: MFCD13560224
CAS Number: 1219980-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR