3-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049821-500MG 049821-2.5G
101889-184EA 364.62 USD
101889-184 101889-186
3-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride
3-[2-(2-Propynyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO
MDL Number: MFCD13560223
CAS Number: 1219949-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR