3-{2-[2-(2-Methoxyethoxy)ethoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049817-500MG 049817-2.5G
101889-168EA 364.62 USD
101889-168 101889-170
3-{2-[2-(2-Methoxyethoxy)ethoxy]ethyl}piperidine hydrochloride
3-{2-[2-(2-Methoxyethoxy)ethoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO₃
MDL Number: MFCD13560219
CAS Number: 1219980-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR