2-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049814-500MG 049814-2.5G
101889-156EA 364.62 USD
101889-156 101889-158
2-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride
2-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄ClNO₂
MDL Number: MFCD13560216
CAS Number: 1220028-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR