3-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049813-500MG
049813-2.5G
101889-152EA
364.62
USD
101889-152
101889-154
3-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride
3-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₄ClNO₂ |
MDL Number:
MFCD13560215 CAS Number: 1220017-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...