4-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049812-500MG 049812-2.5G
101889-148EA 364.62 USD
101889-148 101889-150
4-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride
4-[2-(2-Ethoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄ClNO₂
MDL Number: MFCD13560214
CAS Number: 1219949-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR