2-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049810-500MG 049810-2.5G
101889-140EA 364.62 USD
101889-140 101889-142
2-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride
2-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO₂
MDL Number: MFCD13560212
CAS Number: 1219979-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR