3-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049809-500MG 049809-2.5G
101889-136EA 364.62 USD
101889-136 101889-138
3-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride
3-[2-(2-Methoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO₂
MDL Number: MFCD13560211
CAS Number: 1219967-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR