3-[(2-Phenoxyethoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049807-500MG 049807-2.5G
101889-128EA 364.62 USD
101889-128 101889-130
3-[(2-Phenoxyethoxy)methyl]pyrrolidine hydrochloride
3-[(2-Phenoxyethoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560209
CAS Number: 1220034-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR